2-Methyl-4,6-dimethoxypyrimidine 98%

Lieferant: Apollo Scientific

OR911753-5G
APOSOR911753-5GEA 108 EUR
APOSOR911753-5G
2-Methyl-4,6-dimethoxypyrimidine 98%
2-Methyl-4,6-dimethoxypyrimidine
Formel: C₇H₁₀N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06657567
CAS-Nummer: 13566-48-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR