6-Chloroindole-2-carboxylic acid 98%

Lieferant: Apollo Scientific

OR911762-1G OR911762-25G OR911762-5G
APOSOR911762-1GEA 91.5 EUR
APOSOR911762-1G APOSOR911762-25G APOSOR911762-5G
6-Chloroindole-2-carboxylic acid 98%
6-Chloroindole-2-carboxylic acid
Formel: C₉H₆ClNO₂
Molecular Weight: 195.602 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01863157
CAS-Nummer: 16732-75-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR