6-Chloroindole-2-carboxylic acid 98%
Lieferant: Apollo Scientific
OR911762-1G
OR911762-25G
OR911762-5G
APOSOR911762-1GEA
91.5
EUR
APOSOR911762-1G
APOSOR911762-25G
APOSOR911762-5G
6-Chloroindole-2-carboxylic acid 98%
6-Chloroindole-2-carboxylic acid
Formel:
C₉H₆ClNO₂ Molecular Weight: 195.602 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01863157 CAS-Nummer: 16732-75-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...