6-Methoxyquinolin-8-amine 98%

Lieferant: Apollo Scientific

Synonyme: 8-Amino-6-methoxyquinoline

OR911787-5G
APOSOR911787-5GEA 55.1 EUR
APOSOR911787-5G
6-Methoxyquinolin-8-amine 98%
6-Methoxyquinolin-8-amine
Formel: C₁₀H₁₀N₂O
Molecular Weight: 174.2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00672902
CAS-Nummer: 90-52-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR