3,4-Dimethoxybenzyl chloride 95%

Lieferant: Apollo Scientific

Synonyme: 3,4-Dimethoxybenzyl Chloride

OR911800-5G
APOSOR911800-5GEA 97.3 EUR
APOSOR911800-5G
3,4-Dimethoxybenzyl chloride 95%
3,4-Dimethoxybenzyl chloride
Formel: C₉H₁₁ClO₂
Molecular Weight: 186.635 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00185554
CAS-Nummer: 7306-46-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR