2-tert-Butylanilin 98%
Lieferant: Apollo Scientific
Synonyme:
2-(tert-Butyl)aniline
OR911839-100G
APOSOR911839-100GEA
195
EUR
APOSOR911839-100G
2-tert-Butylanilin 98%
2-tert-Butylanilin
Formel:
C₁₀H₁₅N Molecular Weight: 149.24 g/mol Siedepunkt: 234 Schmelzpunkt: -60 Dichte: 0.9542 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00130023 CAS-Nummer: 6310-21-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...