4-(Methylthio)benzylalkohol 95%

Lieferant: Apollo Scientific

Synonyme: 4-(Methylthio)benzyl Alcohol

OR911843-25G OR911843-5G
APOSOR911843-25GEA 194 EUR
APOSOR911843-25G APOSOR911843-5G
4-(Methylthio)benzylalkohol 95%
4-(Methylthio)benzylalkohol
Formel: C₈H₁₀OS
Lagertemperatur: Raumtemperatur
MDL: MFCD00009706
CAS-Nummer: 3446-90-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR