Methyl 4-methylthiazole-5-carboxylate 95%

Lieferant: Apollo Scientific

OR911845-5G
APOSOR911845-5GEA 65.4 EUR
APOSOR911845-5G
Methyl 4-methylthiazole-5-carboxylate 95%
Methyl-4-methylthiazole-5-carboxylate
Formel: C₆H₇NO₂S
Lagertemperatur: Raumtemperatur
CAS-Nummer: 81569-44-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR