3-Phenoxypropionitril 95%

Lieferant: Apollo Scientific

OR911855-5G
APOSOR911855-5GEA 63.8 EUR
APOSOR911855-5G
3-Phenoxypropionitril 95%
3-Phenoxypropionitril
Formel: C₉H₉NO
Lagertemperatur: Raumtemperatur
MDL: MFCD00013822
CAS-Nummer: 3055-86-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR