Methyl-4-hydroxy-3-methylbenzoate 98%
Lieferant: Apollo Scientific
Synonyme:
Methyl 4-hydroxy-3-methylbenzoate
OR911863-25G
APOSOR911863-25GEA
219
EUR
APOSOR911863-25G
Methyl-4-hydroxy-3-methylbenzoate 98%
Methyl-4-hydroxy-3-methylbenzoate
Formel:
C₉H₁₀O₃ Molecular Weight: 166.18 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06203848 CAS-Nummer: 42113-13-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...