2,2-Dimethylcyclopropancarbonsäure 97%

Lieferant: Apollo Scientific

OR911879-25G
APOSOR911879-25GEA 641 EUR
APOSOR911879-25G
2,2-Dimethylcyclopropancarbonsäure 97%
2,2-Dimethylcyclopropanecarboxylic acid
Formel: C₆H₁₀O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD04972493
CAS-Nummer: 75885-59-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR