4-Ethoxy-3-methoxybenzaldehyd 98%

Lieferant: Apollo Scientific

OR912035-100G
APOSOR912035-100GEA 161 EUR
APOSOR912035-100G
4-Ethoxy-3-methoxybenzaldehyd 98%
4-Ethoxy-3-methoxybenzaldehyde
Formel: C₁₀H₁₂O₃
Molecular Weight: 180.2 g/mol
Schmelzpunkt: 65
Lagertemperatur: Raumtemperatur
MDL: MFCD00016614
CAS-Nummer: 120-25-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR