2,6-Dimethoxy-p-kresol 95%

Lieferant: Apollo Scientific

Synonyme: 3,5-Dimethoxy-4-hydroxytoluol, 2,6-Dimethoxy-4-methylphenol, 4-Methyl-2,6-dimethoxyphenol, 1,3-Dimethoxy-2-hydroxy-5-methylbenzol

OR912037-100G OR912037-25G OR912037-5G
APOSOR912037-100GEA 513 EUR
APOSOR912037-100G APOSOR912037-25G APOSOR912037-5G
2,6-Dimethoxy-p-kresol 95%
2,6-Dimethoxy-p-kresol
Formel: C₉H₁₂O₃
Molecular Weight: 168.19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00017289
CAS-Nummer: 6638-05-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR