2,6-Dimethoxy-p-kresol 95%
Lieferant: Apollo Scientific
Synonyme:
3,5-Dimethoxy-4-hydroxytoluol, 2,6-Dimethoxy-4-methylphenol, 4-Methyl-2,6-dimethoxyphenol, 1,3-Dimethoxy-2-hydroxy-5-methylbenzol
OR912037-100G
OR912037-25G
OR912037-5G
APOSOR912037-100GEA
513
EUR
APOSOR912037-100G
APOSOR912037-25G
APOSOR912037-5G
2,6-Dimethoxy-p-kresol 95%
2,6-Dimethoxy-p-kresol
Formel:
C₉H₁₂O₃ Molecular Weight: 168.19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00017289 CAS-Nummer: 6638-05-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...