(3-Hydroxy-4-methoxyphenyl)essigsäure 95%

Lieferant: Apollo Scientific

Synonyme: 2-(3-Hydroxy-4-methoxyphenyl)acetic acid

OR912038-25G
APOSOR912038-25GEA 95.7 EUR
APOSOR912038-25G
(3-Hydroxy-4-methoxyphenyl)essigsäure 95%
(3-Hydroxy-4-methoxyphenyl)essigsäure
Formel: (C₆H₃(OH)(OCH₃))CH₂COOH
Molecular Weight: 182.18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00016829
CAS-Nummer: 1131-94-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR