5-Bromo-4-chloro-2-methoxyaniline 98%

Lieferant: Apollo Scientific

OR912052-1G
APOSOR912052-1GEA 270 EUR
APOSOR912052-1G
5-Bromo-4-chloro-2-methoxyaniline 98%
5-Bromo-4-chloro-2-methoxyaniline
Formel: C₇H₇BrClNO
Molecular Weight: 236,494 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22192351
CAS-Nummer: 102169-94-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR