4-(4-Oxocyclohexyl)benzonitrile ≥98%

Lieferant: Apollo Scientific

OR912053-5G
APOSOR912053-5GEA 84.1 EUR
APOSOR912053-5G
4-(4-Oxocyclohexyl)benzonitrile ≥98%
4-(4-Oxocyclohexyl)benzonitrile
Formel: C₁₃H₁₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD06410874
CAS-Nummer: 73204-07-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR