1-Butyl-2-pyrrolidone

Lieferant: Apollo Scientific
Achtung

OR912073-25G OR912073-5G
APOSOR912073-25GEA 0 EUR
APOSOR912073-25G APOSOR912073-5G
1-Butyl-2-pyrrolidone
1-Butyl-2-pyrrolidone
Formel: C₈H₁₅NO
Lagertemperatur: Raumtemperatur
MDL: MFCD00020876
CAS-Nummer: 3470-98-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR