2-Amino-2'-methyldiphenyl ether 98%

Lieferant: Apollo Scientific

OR912101-5G
APOSOR912101-5GEA 0 EUR
APOSOR912101-5G
2-Amino-2'-methyldiphenyl ether 98%
2-Amino-2'-methyldiphenyl ether
Formel: C₁₃H₁₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD00025167
CAS-Nummer: 3840-18-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR