5-Ethyl-3-hydroxy-4-methyl-2(5H)-furanon 96%

Lieferant: Apollo Scientific

Synonyme: 5-Ethyl-3-hydroxy-4-methylfuran-2(5H)-one, Ethyl Fenugreek Lactone

OR912126-25G
APOSOR912126-25GEA 85.7 EUR
APOSOR912126-25G
5-Ethyl-3-hydroxy-4-methyl-2(5H)-furanon 96%
5-Ethyl-3-hydroxy-4-methyl-2(5H)-furanon
Formel: C₇H₁₀O₃
Molecular Weight: 142.15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00036673
CAS-Nummer: 698-10-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR