(S)-Methyl-2-hydroxy-3-phenylpropanoate 98%
Lieferant: Apollo Scientific
Synonyme:
(S)-Methyl 2-hydroxy-3-phenylpropanoate, L-β-Phenyllactic acid-OMe
OR912136-100G
APOSOR912136-100GEA
172
EUR
APOSOR912136-100G
(S)-Methyl-2-hydroxy-3-phenylpropanoate 98%
(S)-Methyl-2-hydroxy-3-phenylpropanoate
Formel:
C₁₀H₁₂O₃ Molecular Weight: 180.2005 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00038224 CAS-Nummer: 13673-95-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...