3-Chloro-1,3,4,5-tetrahydro-2H-1-benzazepin-2-one 95%
Lieferant: Apollo Scientific
Synonyme:
3-Chloro-4,5-dihydro-1H-benzo[b]azepin-2(3H)-one
OR912146-5G
APOSOR912146-5GEA
169
EUR
APOSOR912146-5G
3-Chloro-1,3,4,5-tetrahydro-2H-1-benzazepin-2-one 95%
3-Chloro-1,3,4,5-tetrahydro-2H-1-benzazepin-2-one
Formel:
C₁₀H₁₀ClNO Molecular Weight: 195.646 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09260932 CAS-Nummer: 86499-23-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...