1-Fmoc-4-piperidineacetic acid 97%

Lieferant: Apollo Scientific

Synonyme: 2-(1-(((9H-Fluoren-9-yl)methoxy)carbonyl)piperidin-4-yl)acetic acid, Fmoc-4-piperidylacetic acid

OR912176-1G OR912176-5G
APOSOR912176-1GEA 69.6 EUR
APOSOR912176-1G APOSOR912176-5G
1-Fmoc-4-piperidineacetic acid 97%
1-Fmoc-4-piperidineacetic acid
Formel: C₂₂H₂₃NO₄
Molecular Weight: 365.429 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00273478
CAS-Nummer: 180181-05-9

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR