Ethyl-3-cyclopentene-1-carboxylate 95%

Lieferant: Apollo Scientific

Synonyme: Ethyl cyclopent-3-enecarboxylate, 3-Cyclopentene-1-carboxylic Acid Ethyl Ester

OR912181-25G
APOSOR912181-25GEA 174 EUR
APOSOR912181-25G
Ethyl-3-cyclopentene-1-carboxylate 95%
Ethyl-3-cyclopentene-1-carboxylate
Formel: C₈H₁₂O₂
Molecular Weight: 140.18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06410819
CAS-Nummer: 21622-01-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR