5-Amino-2-(methylsulphanyl)benzonitrile 98%

Lieferant: Apollo Scientific

OR912200-1G
APOSOR912200-1GEA 451 EUR
APOSOR912200-1G
5-Amino-2-(methylsulphanyl)benzonitrile 98%
5-Amino-2-(methylsulphanyl)benzonitrile
Formel: C₈H₈N₂S
Molecular Weight: 164.228 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01315282
CAS-Nummer: 306980-70-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR