3-Hydroxy-4-methylbenzonitril 98%

Lieferant: Apollo Scientific

OR912206-25G
APOSOR912206-25GEA 128 EUR
APOSOR912206-25G
3-Hydroxy-4-methylbenzonitril 98%
3-Hydroxy-4-methylbenzonitril
Formel: C₈H₇NO
Molecular Weight: 133.1500 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08361769
CAS-Nummer: 3816-66-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR