1-Butyl-pyrrole-2,5-dione 95%
Lieferant: Apollo Scientific
OR912213-1G
APOSOR912213-1GEA
293
EUR
APOSOR912213-1G
1-Butyl-pyrrole-2,5-dione 95%
1-Butyl-pyrrole-2,5-dione
Formel:
C₈H₁₁NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042756 CAS-Nummer: 2973-09-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...