3-Methyl-1H-indene-2-carboxylic acid 95%
Lieferant: Apollo Scientific
OR912234-1G
APOSOR912234-1GEA
451
EUR
APOSOR912234-1G
3-Methyl-1H-indene-2-carboxylic acid 95%
3-Methyl-1H-indene-2-carboxylic acid
Formel:
C₁₁H₁₀O₂ Molecular Weight: 174,2 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00044428 CAS-Nummer: 34225-81-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...