6-Isopropylchinolin 95%

Lieferant: Apollo Scientific

OR912247-5G
APOSOR912247-5GEA 256 EUR
APOSOR912247-5G
6-Isopropylchinolin 95%
6-Isopropylchinolin
Formel: C₁₂H₁₃N
Molecular Weight: 171,24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00047615
CAS-Nummer: 135-79-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR