1-Chlor-2-ethoxybenzol 95%

Lieferant: Apollo Scientific

Synonyme: 1-Chloro-2-ethoxybenzene, 2-Chlorophenetole

OR912269-5G
APOSOR912269-5GEA 63.8 EUR
APOSOR912269-5G
1-Chlor-2-ethoxybenzol 95%
1-Chloro-2-ethoxybenzene
Formel: C₈H₉ClO
Molecular Weight: 156.61 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00058930
CAS-Nummer: 614-72-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR