trans-4-Butylcyclohexancarbonsäure 98%

Lieferant: Apollo Scientific

OR912272-100G
APOSOR912272-100GEA 66.7 EUR
APOSOR912272-100G
trans-4-Butylcyclohexancarbonsäure 98%
trans-4-Butylcyclohexanecarboxylic acid
Formel: C₁₁H₂₀O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00059560
CAS-Nummer: 38289-28-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR