3-(m-Tolyl)propionsäure 97%

Lieferant: Apollo Scientific

Synonyme: 3-(3-Methylphenyl)propionsäure, 3-(3-Methylphenyl)propansäure

OR912273-5G
APOSOR912273-5GEA 94.4 EUR
APOSOR912273-5G
3-(m-Tolyl)propionsäure 97%
3-(m-Tolyl)propionsäure
Formel: C₁₀H₁₂O₂
Molecular Weight: 164.2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00060326
CAS-Nummer: 3751-48-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR