1-(4-Nitrophenyl)imidazol 95%

Lieferant: Apollo Scientific

Synonyme: 1-(4-Nitrophenyl)-1H-imidazol

OR912274-5G
APOSOR912274-5GEA 59.6 EUR
APOSOR912274-5G
1-(4-Nitrophenyl)imidazol 95%
1-(4-Nitrophenyl)imidazole
Formel: C₉H₇N₃O₂
Molecular Weight: 189.17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00060491
CAS-Nummer: 2301-25-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR