L-Leucic acid 96%

Lieferant: Apollo Scientific

OR912286-100G
APOSOR912286-100GEA 98.6 EUR
APOSOR912286-100G
L-Leucic acid 96%
L-Leucic acid
Formel: C₆H₁₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00064214
CAS-Nummer: 13748-90-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR