N-Acetyl-D-leucin 98%
Lieferant: Apollo Scientific
Synonyme:
N-Acetyle-D-Leucine, N-Ac-D-Leu-OH, Ac-D-Leu-OH, 2-(Acetylamino)-4-methylpentanoic acid
OR912302-100G
APOSOR912302-100GEA
62.5
EUR
APOSOR912302-100G
N-Acetyl-D-leucin 98%
N-Acetyl-D-leucin
Formel:
C₈H₁₅NO₃ Molecular Weight: 173.2 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00066069 CAS-Nummer: 19764-30-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...