3,4-Dichloranisol 97%

Lieferant: Apollo Scientific

OR912314-25G
APOSOR912314-25GEA 62.5 EUR
APOSOR912314-25G
3,4-Dichloranisol 97%
3,4-Dichloranisol
Formel: C₇H₆Cl₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00070785
CAS-Nummer: 36404-30-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR