4-Chloro-3-isopropoxyaniline 97%

Lieferant: Apollo Scientific

OR912325-1G
APOSOR912325-1GEA 364 EUR
APOSOR912325-1G
4-Chloro-3-isopropoxyaniline 97%
4-Chloro-3-isopropoxyaniline
Formel: C₉H₁₂ClNO
Molecular Weight: 185.651 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19624982
CAS-Nummer: 76464-54-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR