3,5-Dimethylbenzaldehyd 98%
Lieferant: Apollo Scientific
OR912342-100G
APOSOR912342-100GEA
203
EUR
APOSOR912342-100G
3,5-Dimethylbenzaldehyd 98%
3,5-Dimethylbenzaldehyde
Formel:
C₉H₁₀O Molecular Weight: 134.18 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00082777 CAS-Nummer: 5779-95-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...