4-Chloro-3-(cyclopropylmethoxy)aniline 95%

Lieferant: Apollo Scientific

OR912358-1G
APOSOR912358-1GEA 632 EUR
APOSOR912358-1G
4-Chloro-3-(cyclopropylmethoxy)aniline 95%
4-Chloro-3-(cyclopropylmethoxy)aniline
Formel: C₁₀H₁₂ClNO
Molecular Weight: 197,661 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19625029
CAS-Nummer: 1265236-35-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR