3-Chloropyridine N-Oxide 98%

Lieferant: Apollo Scientific

Synonyme: 3-Chloropyridine 1-oxide

OR912359-5G
APOSOR912359-5GEA 85.7 EUR
APOSOR912359-5G
3-Chloropyridine N-Oxide 98%
3-Chloropyridine N-Oxide
Formel: C₅H₄ClNO
Molecular Weight: 129.54 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00128857
CAS-Nummer: 1851-22-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR