2-Methyl-2-phenoxypropanoic acid 97%
Lieferant: Apollo Scientific
OR912365-1G
APOSOR912365-1GEA
198
EUR
APOSOR912365-1G
2-Methyl-2-phenoxypropanoic acid 97%
2-Methyl-2-phenoxypropanoic acid
Formel:
C₁₀H₁₂O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00129939 CAS-Nummer: 943-45-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...