1-Ethyl-3-piperidinol 95%
Lieferant: Apollo Scientific
Synonyme:
1-Ethylpiperidin-3-ol, 1-Ethyl-3-piperidinol
OR912385-25G
APOSOR912385-25GEA
59.6
EUR
APOSOR912385-25G
1-Ethyl-3-piperidinol 95%
1-Ethyl-3-hydroxypiperidine
Formel:
C₇H₁₅NO Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006508 CAS-Nummer: 13444-24-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...