trans-4-(Cbz-amino)cyclohexan-1-ol 97%

Lieferant: Apollo Scientific

Synonyme: Benzyl (trans-4-hydroxycyclohexyl)carbamate

OR912410-25G
APOSOR912410-25GEA 127 EUR
APOSOR912410-25G
trans-4-(Cbz-amino)cyclohexan-1-ol 97%
trans-4-(Cbz-amino)cyclohexan-1-ol
Formel: C₁₄H₁₉NO₃
Molecular Weight: 249.306 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00158431
CAS-Nummer: 27489-63-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR