Ethyl-5-formyl-1H-pyrrole-2-carboxylate 95%
Lieferant: Apollo Scientific
Synonyme:
Ethyl 5-formyl-1H-pyrrole-2-carboxylate
OR912411-1G
APOSOR912411-1GEA
71.2
EUR
APOSOR912411-1G
Ethyl-5-formyl-1H-pyrrole-2-carboxylate 95%
Ethyl-5-formyl-1H-pyrrole-2-carboxylate
Formel:
C₈H₉NO₃ Molecular Weight: 167,1600 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00159462 CAS-Nummer: 7126-50-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...