3-Chinolinol 97%

Lieferant: Apollo Scientific

Synonyme: 3-Hydroxyquinoline

OR912414-25G
APOSOR912414-25GEA 114 EUR
APOSOR912414-25G
3-Chinolinol 97%
3-Chinolinol
Formel: C₉H₇NO
Molecular Weight: 145.16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00169018
CAS-Nummer: 580-18-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR