5'-Chlor-2'-methoxyacetophenon 95%

Lieferant: Apollo Scientific

Synonyme: 1-(5-Chloro-2-methoxyphenyl)ethanone

OR912415-25G
APOSOR912415-25GEA 232 EUR
APOSOR912415-25G
5'-Chlor-2'-methoxyacetophenon 95%
5'-Chlor-2'-methoxyacetophenon
Formel: C₉H₉ClO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00174339
CAS-Nummer: 6342-64-9
UN: 1759
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR