N-Cbz-D-phenylalaninol 97%

Lieferant: Apollo Scientific

Synonyme: N-Benzyloxycarbonyl-D-phenylalaninol, Z-D-Phenylalaninol, Cbz-D-Phenylalaninol

OR912423-25G
APOSOR912423-25GEA 105 EUR
APOSOR912423-25G
N-Cbz-D-phenylalaninol 97%
N-Cbz-D-phenylalaninol
Formel: C₁₇H₁₉NO₃
Molecular Weight: 285.3 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00191193
CAS-Nummer: 58917-85-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR