3-(2-Furyl)acrolein 98%

Lieferant: Apollo Scientific

Synonyme: 3-(Furan-2-yl)acrylaldehyde, 3-(2-Furyl)propenal

OR912456-100G
APOSOR912456-100GEA 94.4 EUR
APOSOR912456-100G
3-(2-Furyl)acrolein 98%
3-(2-Furyl)acrolein
Formel: C₇H₆O₂
Molecular Weight: 122.12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00003256
CAS-Nummer: 623-30-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR