4(7)-Nitrobenzimidazole 98%

Lieferant: Apollo Scientific

Synonyme: 7-Nitro-1H-benzo[d]imidazole

OR912459-1G
APOSOR912459-1GEA 388 EUR
APOSOR912459-1G
4(7)-Nitrobenzimidazole 98%
4(7)-Nitrobenzimidazole
Formel: C₇H₅N₃O₂
Molecular Weight: 163,134 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00220143
CAS-Nummer: 10597-52-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR