4-Hydroxy-2-methyl-pyrimidine-5-carboxylic acid ethyl ester 95%

Lieferant: Apollo Scientific

OR912465-100G OR912465-25G
APOSOR912465-100GEA 951 EUR
APOSOR912465-100G APOSOR912465-25G
4-Hydroxy-2-methyl-pyrimidine-5-carboxylic acid ethyl ester 95%
4-Hydroxy-2-methyl-pyrimidine-5-carboxylic acid ethyl ester
Formel: C₈H₁₀N₂O₃
Molecular Weight: 182.177 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00223606
CAS-Nummer: 67383-32-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR